Learn More
Biodesign™ Leveling Platform G205
Brand:  Biodesign™ G205
Product Code. 12876512
  • 1400.00 DKK / Each
Only null left
Estimated Shipment: 05-09-2025
Description
10mm thick acrylic top with three individually adjustable chrome plated metal feet and a large easy to read bulls-eye level on the top. Ensures a level surface when pouring and running polyacrylamide and agarose gels.
Specifications
Specifications
Provides a level surface for pouring electrophoresis gels
Three individually adjustable chrome-plated metal feet and a large easy-to-read bulls-eye level
0.4 in.
28 x 28 x 1 cm
28 cm
28 cm
Leveling Platform
Acrlyic
1 cm
11 in.
11 in.
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Biodesign™ Leveling Platform G205 > Dimensions: 28 x 28cm