missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Lid for 300 L, 310 L, 480 L, and 500 L Large Volume Containers

Lid for 300 L, 310 L, and 500 L double-wall and large volume containers, and 480 L pallet container.

Brand:  Gilac™ G156877

Product Code. 17938421

  • 1635.00 DKK / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Lid for 300 L, 310 L and 500 L double-wall and large volume containers and 480 L pallet container.
Specifications

Specifications

Bin Lid
HDPE
Gray
Rectangular
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers