Learn More
LIFR alpha Antibody (12D3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Bio-Techne NBP3-18054
Product Code. 18310474
  • 2219.16 DKK / 0.10mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
LIFR alpha Monoclonal antibody specifically detects LIFR alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
Specifications
LIFR alpha
Monoclonal
1 mg/ml
Western Blot, Flow Cytometry 2-5 ug/ml, ELISA
CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS
Human recombinant soluble LIFR alpha-Fc
0.1 mg
Immunology
3977
Store at 4C. Do not freeze.
IgG1 κ
Western Blot, Flow Cytometry, ELISA
12D3
Unconjugated
PBS, pH 7.4
Mouse
Protein A purified
RUO
Primary
Human
Purified