missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN-28A Rabbit anti-Human, Mouse, Rat, Clone: 2S1I10, Novus Biologicals™
Rabbit Monoclonal Antibody
1460.01 DKK - 3692.13 DKK
Specifications
Antigen | LIN-28A |
---|---|
Clone | 2S1I10 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18310863
|
Novus Biologicals
NBP3-16843-20UL |
20 μg |
1460.01 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18392745
|
Bio-Techne
NBP3-16843-100UL |
100 μg |
3692.13 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIN-28A Monoclonal antibody specifically detects LIN-28A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
LIN-28A | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Epigenetics, Neuroscience, Stem Cell Markers, Stem Cells | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
79727 | |
IgG | |
Affinity purified |
2S1I10 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CSDD1lin-28A, FLJ12457, LIN-28, lin-28 homolog (C. elegans), lin-28 homolog A (C. elegans), Lin-28A, LIN28RNA-binding protein LIN-28, ZCCHC1protein lin-28 homolog A, Zinc finger CCHC domain-containing protein 1, zinc finger, CCHC domain containing 1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human LIN-28A (Q9H9Z2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |