Learn More
MP Biomedicals™ Linbro™ Liquisystem, Polypropylene Tubes
Used as Lab essentials
Brand: MP Biomedicals™ 0961226C2
704.90 DKK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
This item is not returnable.
View return policy
Description
- Storage System
- Sterile
- Store at room temperature
Specifications
Each | |
Polypropylene | |
Sterile |
Linbro Liquisystem | |
2°C to 8°C |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
39Hours
17Minutes
3Seconds
39
Product Content Correction
Product Title
MP Biomedicals™ Linbro™ Liquisystem, Polypropylene Tubes > Linbro Liquisystem
Spot an opportunity for improvement?Share a Content Correction