Learn More
Thermo Scientific™ Locator™ and Locator Plus System Carts
Effortlessly move Thermo Scientific™ Locator system vessels when using Locator and Locator Plus System Carts.
Brand: Thermo Scientific™ AY509X1-70
3357.75 DKK valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
Effortlessly move rack and box systems around the lab using Thermo Scientific™ Locator™ and Locator Plus System Carts.
With wheels. Molded to fit snugly around vessel base.
Specifications
Locator 6 Plus cart | |
Locator 6 Plus Vessel |
Promotions
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now