missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Lomustine Product Code.: 16617032

Thermo Scientific Chemicals Lomustine

Product Code. 16617032
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 16617032

Brand: Thermo Scientific Chemicals 465490010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13010-47-4
Molecular Formula C9H16ClN3O2
Molecular Weight (g/mol) 233.70
MDL Number MFCD00012392
InChI Key GQYIWUVLTXOXAJ-UHFFFAOYSA-N
IUPAC Name 3-(2-chloroethyl)-1-cyclohexyl-3-nitrosourea
SMILES ClCCN(N=O)C(=O)NC1CCCCC1

Specifications

Alpha Vector LOMUSTINE
Melting Point 89°C to 93°C
Color Yellow
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 1 g
UN Number 2811
Sensitivity Light sensitive
Solubility Information soluble aq. soln., soluble in acetone, chloroform, ethanol
Formula Weight 233.7
Physical Form Powder
Chemical Name or Material Lomustine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467268 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number465490010,465492500
Lot NumberA0443839 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number465490010,465492500
Lot NumberA0457798 Certificate TypeCertificate of Analysis Date23/11/2023 Catalog Number465490010,465492500
Lot NumberA0450457 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number465490010,465492500
Lot NumberA0416119 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number465490010,465492500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Lomustine >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.