Learn More
Abnova™ LongTaq DNA Polymerase
LongTaq DNA polymerase catalyzes 5'-3' synthesis of DNA.
Brand: Abnova™ D0013.1000Units
This item is not returnable.
View return policy
Description
- The enzyme has been proved not having the 3′-5′ exonuclease activity.
- The enzyme has proven to have high amplification yield, be stable at high temparetaure.
Specifications
Store at -20°C Guaranteed stable for 6 months when properly stored | |
Standard PCR | |
20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol |
1000 units | |
5 units/μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ LongTaq DNA Polymerase > 1000 units
Spot an opportunity for improvement?Share a Content Correction