Learn More
Abnova™ LongTaq DNA Polymerase
LongTaq DNA polymerase catalyzes 5'-3' synthesis of DNA.
Brand:  Abnova™ D0013.2500Units
 View more versions of this product
Product Code. 16062555
  • 11110.00 DKK / 2500 units
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • The enzyme has been proved not having the 3′-5′ exonuclease activity.
  • The enzyme has proven to have high amplification yield, be stable at high temparetaure.
Specifications
Specifications
Store at -20°C Guaranteed stable for 6 months when properly stored
Standard PCR
20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol
2500 units
5 units/μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ LongTaq DNA Polymerase > 2500 units