Learn More
Lonza™ BioWhittaker™ 293 Cell Serum-Free Media
Completely balanced to support vaccine and viral vector production of high-density human embryonic kidney (HEK) 293 cell culture environments
Brand: Lonza™ BE12-764Q
This item is not returnable.
View return policy
Description
Very low levels of recombinant proteins facilitate downstream purification and provide regulatory compliance
- Chemically-defined media optimized for specific cell types
† Store as detailed on label.
Specifications
Pro293A-CDM | |
For Adherent Cells | |
Bottle |
No Glutamine, No Phenol Red | |
1 L | |
2° to 8°C |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lonza™ BioWhittaker™ 293 Cell Serum-Free Media > Pro293a-cdm; for adherent 293 cells
Spot an opportunity for improvement?Share a Content Correction