missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ BioWhittaker™ Ham's F-10 Medium

Brand:  Lonza™ BE12-618F

Product Code. 11665200

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Originally designed for low density, serum-free growth of Chinese Hamster Ovary (CHO) cells
  • Also used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2° to 8°C
Sterile
500 mL
Ham's F10
Sterile-filtered through two consecutive 0.2 μm filters
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lonza™ BioWhittaker™ Ham's F-10 Medium > Ham's F-10; 500ml

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.