missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ BioWhittaker™ McCoy's 5A Medium (Modified)

General purpose medium for primary and established cell lines. BioWhittaker™ McCoy's 5A Medium (Modified) is manufactured in accordance with cGMP regulations.

Brand:  Lonza™ BE12-688F

 View more versions of this product

Product Code. 11645210

This item is not returnable. View return policy

Description

Description

  • Formulated for the nutritional requirements of Walker 256 carcinoma
  • Unique components include asparagine, 3X normal glucose, amino acids (all L-isomers) and increased folic acid
  • Manufactured in accordance with cGMP regulations
  • Formulations used for standard liquid media products are those recommended by Tissue Culture Association
TRUSTED_SUSTAINABILITY
Specifications

Specifications

McCoy's 5A
Sterile-filtered through two consecutive 0.2 μm filters
6.89 to 7.2
Sterile
500 mL
2° to 8°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lonza™ BioWhittaker™ McCoy's 5A Medium (Modified) > w/ L-Glutamine; Quantity: 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.