missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ DNA Ladders
Each band has 50% GC content for precise sizing and sharp resolution
2060.40 DKK - 2470.00 DKK
Specifications
Product Type | DNA Molecular Weight Marker/Ladder |
---|---|
Form | Liquid |
Formulation | Supplied at various concentrations in TE Buffer and each includes a vial of 6X Sample Loading Buffer 1mL total volume 15% w/v Ficoll 400, 0.06% w/v Bromophenol Blue, 0.06% w/v Xylene cyanol FF, 30mM EDTA |
For Use With (Application) | Size determination of DNA samples |
Storage Requirements | 2° to 8°C |
Product Code | Brand | Quantity | No. of Bands | Concentration | Increments | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | No. of Bands | Concentration | Increments | Price | Quantity & Availability | ||||||
11560777
|
Lonza™
50320 |
150 μL | 50 | 200 ng/μL | 20bp | N/A | N/A | N/A | |||||
11580777
|
Lonza™
50322 |
150 μL | 30 | 200 ng/μL | 100bp |
2470.00 DKK
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11570777
|
Lonza™
50321 |
160 μL | 10 | 100 ng/μL | 100bp | N/A | N/A | N/A | |||||
11590777
|
Lonza™
50323 |
300 μL | 16 | 200 ng/μL | 500bp |
2470.00 DKK
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11540787
|
Lonza™
50330 |
150 μL | 25 | 200 ng/μL | 20bp |
2470.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Specifications
DNA Molecular Weight Marker/Ladder | |
Supplied at various concentrations in TE Buffer and each includes a vial of 6X Sample Loading Buffer 1mL total volume 15% w/v Ficoll 400, 0.06% w/v Bromophenol Blue, 0.06% w/v Xylene cyanol FF, 30mM EDTA | |
2° to 8°C |
Liquid | |
Size determination of DNA samples |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lonza™ DNA Ladders