missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ DNA Standards for Pulsed Field Gel Electrophoresis
For size estimation of megabase DNA separated by pulsed field gel electrophoresis (PFGE)
4115.00 DKK
Specifications
Product Type | DNA Molecular Weight Marker/Ladder |
---|---|
For Use With (Application) | Size estimation of megabase DNA separated by pulsed field Gel electrophoresis |
Quantity | 5 Plugs |
Storage Requirements | 2° to 8°C |
Product Code | Brand | Formulation | No. of Bands | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | No. of Bands | Price | Quantity & Availability | ||||
11520797
|
Lonza™
50401 |
lambda ladder, 50mM EDTA, 10mM [Tris(hydroxymethyl)aminoethane], pH 8, 5μg/mL Proteinase K 0.01% N-lauroyl Sarcosine | 18 |
4115.00 DKK
Pack of 5 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
||||
Description
- Ladders are prepared from five highly purified DNA fragments in perfect relative mass equivalence
- Reverse QuantLadders reverse the mass equivalence order of the QuantLadder
- SimplyLoad™ ready-to-load ladders contain loading buffer; other ladders are supplied with separate loading buffer
- Store at 2° to 8°C for up to 12 months
Specifications
DNA Molecular Weight Marker/Ladder | |
Size estimation of megabase DNA separated by pulsed field Gel electrophoresis | |
5 Plugs | |
2° to 8°C |