Learn More
3M™ Particulate Respirator, Series C100
Click to view available options
Filter Type:
FFP1
FFP2
With Exhalation Valve:
No
Yes
Unit Size:
Pack of 10
Pack of 20
Description
Provides cost-effective, lightweight, comfortable, and effective protection against dust and mist.
- The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes
- Exhalation valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions
Specifications
Specifications
Filter Type | FFP1 |
Material | Polypropylene |
Product Type | General Purpose Particulate Respirator |
With Exhalation Valve | No |
Model | C101 |
Sterility | Non-sterile |
FDA Cleared Surgical Mask | No |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
3M™ Particulate Respirator, Series C100 >
Spot an opportunity for improvement?Share a Content Correction