Learn More
SI Analytics™ Low Maintenance pH Combination Electrode with Screw Plug Head
Ensure high measurement accuracy, stability, and long service life while maintaining high adaptability to your measurement tasks. SI Analytics™ Low Maintenance pH Combination Electrode with Screw Plug Head is robust, durable, and easy to clean.
Brand:  SI Analytics™ 285111015
2253.21 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10490602
  • 2480.00 DKK / Each
Only null left
Estimated Shipment: 18-08-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • PH combination electrode, type H8281HD, length 120 mm, D = 12 mm, 0 ... + 100 ° C
  • Low Maintenance pH Combination Electrode with Screw Plug Head
  • Low maintenance, glass shaft, KPG annular gap junction
  • Referid™-electrolyte, Silamid™-reference system, dome membrane, H-glass, plug head, length 120 mm, 12 mm Ø, 0...+100 °C, 2...13 pH
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Screw Plug Head
H-Glass
Low Maintenance pH Combination Electrode
Dome
0°C to 100°C
12 mm
KPG Annular Gap Diaphragm
2 to 13
Silamid™
H 8281 Hd
120 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Manuals
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SI Analytics™ Low Maintenance pH Combination Electrode with Screw Plug Head > LengthMetric: 120mm