Learn More
LP-533401 hydrochloride, MedChemExpress
MedChemExpress LP-533401 hydrochloride is a tryptophan hydroxylase 1 inhibitor that regulates serotonin production in the gut.
Brand: Medchem Express HY-15849A
This item is not returnable.
View return policy
Description
LP-533401 hydrochloride is a tryptophan hydroxylase 1 inhibitor that regulates serotonin production in the gut.Specifications
LP-533401 hydrochloride | |
Light Yellow | |
C27H23ClF4N4O3 | |
DMSO : 25 mg/mL (44.41 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) | |
562.94 | |
98.62% | |
Solid |
1040526-12-2 | |
Research | |
5 mg | |
N[C@@H](CC1=CC=C(C2=NC(N)=NC(OC(C3=CC=C(C4=CC=CC(F)=C4)C=C3)C(F)(F)F)=C2)C=C1)C(O)=O.[H]Cl | |
562.94 | |
Research | |
COVID-19-immunoregulation |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
LP-533401 hydrochloride, MedChemExpress