Learn More
LP-533401 hydrochloride, MedChemExpress
MedChemExpress LP-533401 hydrochloride is a tryptophan hydroxylase 1 inhibitor that regulates serotonin production in the gut.
Brand:  Medchem Express HY-15849A
Encompass_Preferred
 View more versions of this product
Product Code. 16479407
  • 1037.34 DKK / 5mg
Explore more special offers
Description
Description
LP-533401 hydrochloride is a tryptophan hydroxylase 1 inhibitor that regulates serotonin production in the gut.
Specifications
Specifications
LP-533401 hydrochloride
Light Yellow
C27H23ClF4N4O3
DMSO : 25 mg/mL (44.41 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
562.94
98.62%
Solid
1040526-12-2
Research
5 mg
N[C@@H](CC1=CC=C(C2=NC(N)=NC(OC(C3=CC=C(C4=CC=CC(F)=C4)C=C3)C(F)(F)F)=C2)C=C1)C(O)=O.[H]Cl
562.94
Research
COVID-19-immunoregulation
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
LP-533401 hydrochloride, MedChemExpress