missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP-6, p Ser1490 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-12921
This item is not returnable.
View return policy
Description
LRP-6 Polyclonal antibody specifically detects LRP-6 in Human samples. It is validated for Western BlotSpecifications
LRP-6 | |
Polyclonal | |
PBS, 1% BSA, 20% Glycerol (pH7) | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
ADCAD2, FLJ90062, FLJ90421, low density lipoprotein receptor-related protein 6, low-density lipoprotein receptor-related protein 6, LRP-6 | |
Carrier-protein conjugated synthetic peptide corresponding to residues around human LRP-6 (phospho Ser1490). The exact sequence is proprietary. | |
100 μg | |
Phospho Specific, Stem Cell Signaling Pathway | |
4040 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |