Learn More
Corning™ LSE Digital Microplate Shaker
Large capacity – process up to 4 microplates
Brand:  Corning™ 6782-4
6922.91 DKK valid until 2025-04-30
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 11971991
  • 8390.58 DKK / Each
Estimated Shipment: 10-09-2025
to see stock.
Explore more special offers
Description
The Corning LSE digital microplate shaker features digital control of shaking speed and run time. A single control knob is used to set parameters, and values are shown on a large 3-digit display. A sturdy base encloses the motor for quiet, vibration-free operation.
The included stainless steel platform accommodates up to 4 microplates (standard or deep well), and the shaker is safe for use in temperature controlled environments from 4°C to 65°C.
  • Large capacity – process up to 4 microplates
  • 3 mm orbit for resuspending, homogenizing, or vortexing
  • Large digital display with single-knob control of speed time
  • Suitable for cold room or incubator use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6782-4
Product Title
Corning™ LSE Digital Microplate Shaker > Model: 6782-4