Learn More
Invitrogen™ Luteinizing Hormone Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHLHX10
This item is not returnable.
View return policy
Description
LH is a member of the glycoprotein hormone family, Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. LH is expressed in the pituitary gland and promotes spermatogenesis and ovulation by stimulating the testes and ovaries to synthesize steroids. The genes for the beta chains of chorionic gonadotropin and for luteinizing hormone are contiguous on chromosome 19q13.3. Mutations in this gene are associated with hypogonadism which is characterized by infertility and pseudohermaphroditism.Specifications
P01215, P01229 | |
140 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Cga, LHB | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
156.3 to 10,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
1081, 3972 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 50 μL|Serum, 50 μL|Supernatant, 100 μL | |
Luteinizing Hormone | |
4 hr 45 min |