Learn More
Lutetium pieces, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11306607
Lutetium pieces, 99.9% (REO), Thermo Scientific Chemicals
Product Code. 11306607
5 g
Click to view available options
Quantity:
1 g
5 g
Product Code. 11306607
Brand: Thermo Scientific Alfa Aesar 000628.06
Request Bulk or Custom Format
CAS: 7439-94-3 | Lu | 174.97 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged in air-stable glass bottle
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7439-94-3
Molecular Formula Lu
Molecular Weight (g/mol) 174.97
MDL Number MFCD00011098
InChI Key OHSVLFRHMCKCQY-UHFFFAOYSA-N
Synonym lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces
PubChem CID 23929
ChEBI CHEBI:33382
IUPAC Name lutetium
SMILES [Lu]
Specifications
Quantity 5 g
Percent Purity 99.9%
Odor Odorless
Packaging Glass bottle
Form Pieces
Assay (REO)
Chemical Name or Material Lutetium pieces
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM16L013 Certificate TypeCertificate of Analysis Date17/01/2025 Catalog Number000628.06,000628.03
Lot NumberZ24K006 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number000628.06,000628.03
Lot NumberW13K016 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog Number000628.06,000628.03
Lot NumberS14K023 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog Number000628.06,000628.03
Lot NumberU22J002 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog Number000628.06,000628.03
RUO – Research Use Only
Product Content Correction
Product Title
Lutetium pieces, 99.9% (REO), Thermo Scientific Chemicals >