Learn More
Thermo Scientific™ Magnesium Oxide Crucible, Cylindrical, Flat Base;OD (mm), 32;Height (mm), 32
Brand: Thermo Scientific™ 045854.KS
1119.14 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
32 mm | |
Magnesium Oxide | |
1 Each |
32 mm | |
Crucible | |
Cylindrical |