Learn More
Magnesium sulfate heptahydrate, 99.5%, for analysis
CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol
378.00 DKK - 5665.00 DKK
Chemical Identifiers
CAS | 10034-99-8 |
---|---|
Molecular Formula | H14MgO11S |
Molecular Weight (g/mol) | 246.47 |
MDL Number | MFCD00149785 |
InChI Key | WRUGWIBCXHJTDG-UHFFFAOYSA-L |
Synonym | magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate |
PubChem CID | 24843 |
ChEBI | CHEBI:31795 |
IUPAC Name | magnesium;sulfate;heptahydrate |
SMILES | O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10256840
|
Thermo Scientific Acros
213115000 |
500 g | Plastic bottle |
378.00 DKK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10135453
|
Thermo Scientific Acros
213110025 |
2.5 kg | Plastic bottle |
1110.00 DKK
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10378703
|
Thermo Scientific Acros
213110250 |
25 kg | Fibre drum |
5665.00 DKK
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10034-99-8 | |
246.47 | |
WRUGWIBCXHJTDG-UHFFFAOYSA-L | |
24843 | |
magnesium;sulfate;heptahydrate |
H14MgO11S | |
MFCD00149785 | |
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate | |
CHEBI:31795 | |
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O |
Specifications
770.0°C | |
White | |
Crystals | |
H14MgO11S | |
MFCD00149785 | |
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate | |
WRUGWIBCXHJTDG-UHFFFAOYSA-L | |
magnesium;sulfate;heptahydrate | |
24843 | |
246.48 | |
Analytical |
10034-99-8 | |
5 to 8 (5% soln. at 25°C) | |
99.4% min. (Complexometry) | |
MgSO4·7H2O | |
15, 5755 | |
Solubility in water: 710g/L (20°C). Other solubilities: soluble in glycerol, sparingly soluble in alcohol | |
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O | |
246.47 | |
CHEBI:31795 | |
99.5% | |
Magnesium sulfate heptahydrate, p.a. |
Safety and Handling
missing translation for 'rtecsNumber' : OM4508000
- Magnesium sulfate heptahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.