Specifications
ISO 7010 | |
Plastic | |
Permanent Acrylic | |
297 mm | |
Diamond | |
Transport Sign |
ADR 3A-297*297-B0859 | |
1 Ea. | |
Magnetic Plastic | |
297 mm | |
Highly Flammable Liquid 3 | |
Red |