Learn More
Velp Scientifica™ Magnetic Stirring Bars
Emerse this into a liquid to produce a stirring movement. Velp Scientifica™ Magnetic Stirring Bars do not contaminate or react with the mixure they are in.
Brand:  Velp Scientifica™ A00001062
 View more versions of this product
Product Code. 15329514
  • 23.00 DKK / Each
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Description
Emerse this into a liquid to produce a stirring movement. Velp Scientifica™ Magnetic Stirring Bars do not contaminate or react with the mixure they are in.
  • For stirring applications
  • Chemically inert
Specifications
Specifications
VELP Heating magnetic stirrer with round top
3 mm
6 mm
Stir Bar