Learn More
3M™ Magnifying Lens
3M™ Magnifying Lens is used for magnifying object and can be used with all Speedglas welding helmets.
229.00 DKK
Specifications
For Use With (Equipment) Speedglas
Product Type Magnifying Lens
View More Specs
Products 2
Product Code Brand Power Intensity Price Quantity & Availability  
Product Code Brand Power Intensity Price Quantity & Availability  
15879644
View Documents
3M™
7000144517
2.5X
229.00 DKK
Each
Estimated Shipment: 04-08-2025
to see stock.
Add to basket
15869634
View Documents
3M™
7000028023
2.0X
229.00 DKK
Each
Estimated Shipment: 04-08-2025
to see stock.
Add to basket
Description
Description
  • On the 3M™ Speedglas™ Welding Helmet Series 9100 there is a magnifying lens holder built into the welding filter, making lens installation easier and more secure
  • For the new 3M™ Speedglas™ Welding Helmet Series 100 there is a magnifying lens holder built into the welding filter, making lens installation easier and more secure. For the older 100 series filters you will need a holder for the magnifier
  • For the 3M™ Speedglas™ Welding Helmet SL you will need to add the holder for magnifier as a separate spare part (171031)
Specifications
Specifications
Speedglas
Magnifying Lens
Documents
Product Certifications
Product Content Correction
Product Title
3M™ Magnifying Lens