missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., 99.0-101.0%, Honeywell Fluka™

Product Code. 15623400
Click to view available options
Quantity:
500 g
1 kg
50 kg
6 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur., 99.0-101.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+);sulfate;hydrate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100.0000%
Quantity 50 kg
Assay Percent Range 99.0 to 101.0%
Linear Formula CuSO4 · 5H2O
UN Number UN3077
Formula Weight 169.02
Percent Purity 99.0 to 101.0%
Grade ACS Reagent/Reagent Ph. Eur.
Packaging Fiberboard Box
Density 2.950 g/cm3
Chemical Name or Material Manganese(II) sulfate monohydrate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., 99.0-101.0%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.