Learn More
BUCHI Manifold Ampoule Adapter
Includes 19 ampoule connections and cap adaptor.
Brand: BUCHI 11065725
This item is not returnable.
View return policy
Description
- Includes 19 ampoule connections and a cap adapter
Specifications
Ampule Adapter | |
Ambient to -105°C | |
L-200 Freeze Dryer | |
5.9 in. | |
3.9 in. | |
2.4 lb. |
Variable | |
Rubber | |
110 mm | |
150 mm | |
100 mm | |
1.1 kg |