Learn More
Scotsman™ Ice Flaker with Storage Bin
Extrudes ice flakes at a temperature just below zero degrees. Scotsman™ Ice Flaker with Storage bin can produce up to 70kg of flake per day and holds up to 25kg of storage. Flakes contain 25% residual water content and is stored inside an insulated, plastic lined containment bin with front opening for easy access.
Brand:  Scotsman™ AF80
Product Code. 12338697
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
Ice flakes are extruded at a temperature just below zero degrees and contain 25% residual water content. The flake ice is stored inside an insulated, plastic lined storage bin with front opening for easy access. Electrical supply 230V/50Hz.
  • Up to 70kg (154 lbs.) of flake ice production per day
  • Up to 25kg (55 lbs.) built-in ice storage
  • Pressure-based water level monitoring system may work with demineralised water as well as with standard tap water
  • Electronically controlled operation, with self-diagnostic functions with external alarm lights providing operational information for early alert and fast diagnosis of operating issues
  • Plug-n-play configuration right out of the box - just connect incoming water line and water discharge
  • External components constructed from corrosion-resistant stainless steel, crafted for optimal aesthetic appeal through superior fit and finish
  • Ergonomically convenient, slanted & disappearing storage bin door
  • Front access for easier maintenance
  • Front-in, front-out airflow allows for built-in installations
  • CFC-free R-134a refrigerant
  • 120mm legs included, extensions available as an option Scotsman flaked ice is ideal for a variety of applications within laboratory and industrial environments.
Specifications
Specifications
ISO 9001
AF80 Ice maker
120mm legs
Corrosion-resistant stainless steel
230V
70kg/day
2 year manufacturer warranty
626mm
May work with demineralised water as well as with standard water
50Hz
813mm
535mm
230V/50Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Scotsman™ Ice Flaker with Storage Bin > Capacity: 25kg