Learn More
Socorex™ Acura Manual 810 Dilution Micropipettes
Use graduated glass pipettes when performing serial 1:10 dilutions in bacteriology with Socorex™ Acura Manual 810 Dilution Micropipettes. They feature smooth activation with excellent ergonomics.
Brand:  Socorex™ 810.1100
1715.15 DKK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Product Code. 10337803
  • 2305.00 DKK / Each
The Ultimate Lab Appliances Promotion
Use promo code 24075 at checkout
View Available Promotions
Only null left
Estimated Shipment: 05-08-2025
Description
  • Three-year warranty
  • No pre-calibrated fixed volumes
  • No setting required
  • Independent calibration for each volume
  • Autoclavable
Specifications
Specifications
CE Marked, IVD
Manual 810 Dilution
Dilution Pipette
1 Each
Autoclavable
Acura
1 +0.1 mL
Certificates
View accessory products
Promotions
Promotions
  • Multiple Manufacturers
    Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.
    Promo Code: 24075
    Expires2025-06-30
    Days
    12
    Hours
    19
    Minutes
    34
    Seconds
    0
Product Content Correction
Product Title
Socorex™ Acura Manual 810 Dilution Micropipettes > Certification: CE Marked, IVD