Learn More
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Brand: Thermo Scientific™ C4030-100
2815.02 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Lightweight, precision crimpers attach seals to chromatography or serum vials.- Factory set to propery crimp standard-thickness seals
- Adjustable handle setting scree allows proper crimps with septa of varying thickness
Specifications
Manual Crimper, Standard | |
National Vial, Chromacol Vial | |
Industrial Chromatography |
Manual Crimper | |
30 mm | |
National |