Learn More
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Brand:  Thermo Scientific™ C4028-102
2849.79 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12852344
  • 3005.00 DKK / Each
Description
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Place decrimping head squarely on seal and squeeze.
  • Minimizes damage to vial rim with chromatography or serum vials 
  • Removes standard aluminum seals and many steel seals
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Manual Decrimper
Manual Decrimper
28 mm
National
1 Ea.
Aluminum And Steel Seal, National Vial, Chromacol Vial
Industrial Chromatography
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers > Decrimper, For 28mm crimp seals