Learn More
Fisherbrand™ Manual Decapper, Steel, Non-autoclavable
Manual decrimping tools allow easy removal of aluminum seals without breakage.
Brand: Fisherbrand™ 08 07 0001
2949.75 DKK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
- Decapper
- Manual
- Steel
Specifications
Aluminum Cap | |
8mm Decapper |