missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MAPK14 (p38 alpha), GST Tag Recombinant Protein
Human MAPK14 (p38 alpha), GST Tag Recombinant Protein
4680.00 DKK - 59400.00 DKK
Specifications
Accession Number | NP_620581 |
---|---|
Species | Human |
Protein Tag | GST Tag |
Expression System | E. coli |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10083242
|
Invitrogen
PV3598 |
1 mg |
59400.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11573417
|
Invitrogen
PV3304 |
10 μg |
4680.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAPK14 (p38 alpha) is a member of the stress-activated protein kinase class of MAPKs.Specifications
NP_620581 | |
Human | |
GST Tag | |
E. coli |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human MAPK14 (p38 alpha), GST Tag Recombinant Protein