Learn More
Masterflex™ Chem-Durance™ Bio L/S™ High-Performance Precision Pump Tubing
Use this tubing for a wide range of laboratory, pharmaceutical processing and biotech applications. Masterflex™ Chem-Durance™ Bio L/S™ High Performance Precision Pump Tubing features excellent chemical resistance and durability under pressure.
3650.00 DKK - 6200.00 DKK
Specifications
Autoclavable Autoclavable
Material Chem-Durance Bio
View More Specs
Products 4
Product Code Brand Tubing Size Price Quantity & Availability  
Product Code Brand Tubing Size Price Quantity & Availability  
11816953
View Documents
Masterflex™
06442-15
15
3650.00 DKK
Each
Only null left
Estimated Shipment: 30-06-2025
Add to basket
11826953
View Documents
Masterflex™
06442-24
24
4945.00 DKK
Each
Only null left
Estimated Shipment: 11-08-2025
Add to basket
11836953
View Documents
Masterflex™
06442-35
35
6200.00 DKK
Each
Only null left
Estimated Shipment: 31-07-2025
Add to basket
11785648
View Documents
Masterflex™
06442-36
36
6175.00 DKK
Each
Only null left
Estimated Shipment: 08-08-2025
Add to basket
Description
Description
  • For applications involving pressure, suction lift, viscous fluids or long tubing life
  • Thicker wall compared to precision pump tubing
  • Performs in a wide range of chemical applications limiting the number of tubing formulations you need to keep on hand
  • Low spallation maintains purity of fluid
  • Flexible outer jacket acts as an insulating layer to absorb and dissipate heat from the rollers and extending the life of the tubing
  • Plasticizer-free, smooth bore liner is hydrophobic and resists the absorption/adsorption of aqueous fluids Complies with USP Class VI, FDA, REACH, RoHS
  • Excellent chemical compatibility and biocompatibility
  • Excellent pump life and durability under pressure
  • Excellent chemical resistance and long pumping life
  • Meets USP Class VI requirements
Specifications
Specifications
Autoclavable
Chem-Durance Bio
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Masterflex™ Chem-Durance™ Bio L/S™ High-Performance Precision Pump Tubing