Learn More
Masterflex™ L/S™ Easy-Load™ II Pump Head Mounting Hardware
Mount up to four pump heads on a single drive. Masterflex™ Stainless Steel Mounting Hardware is designed for use with L/S™ Easy-Load™ II Pump Heads.
Brand: Masterflex™ 77200-04
634.94 DKK valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
Mount up to four pump heads on a single drive. Masterflex™ Stainless Steel Mounting Hardware is designed for use with L/S™ Easy-Load™ II Pump Heads.
- For use with L/S Easy-Load II Pump Heads
- See individual drive specifications for maximum number of heads to be mounted
- Mounting hardware is stainless steel
Specifications
Mounting four L/S Easy-Load II pump heads | |
1 Each | |
Stainless Steel |
L/S Easy-Load II pump heads | |
Mounting Hardware |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
8Hours
10Minutes
9Seconds
9