Learn More
Thermo Scientific™ ART™ Pipette Tips, 10ul, Sterile Racked, Barrier, 960/pk.
ART Barrier Pipette Tips
Brand: Thermo Scientific™ 2139
604.19 DKK valid until 2025-06-30
Use promo code "25076" to get your promotional price.
Description
Specifications
ART Pipette Tips, 10ul, Sterile | |
Pipette Tips, barrier, ultra micro, sterile | |
Rack, 960 |
Sterile | |
10 ÎĽL | |
Pk of 960 |
Promotions
-
Thermo Scientific ClipTip
,
Thermo Scientific ART
,
Thermo Scientific Finnpipette
,
Thermo Scientific Matrix
Exclusive Promotion on Liquid Handling
Unlock Precision: Top Pipettes and Tips on Promotion!Promo Code: 25076Expires: 2025-06-30
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59% -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
New Offer Save Now -
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30
New Offer Save Up to 78%