Learn More
3M™ Maxim™ DX Safety Glasses
3M™ Maxim™ DX Safety Glasses feature superior eye coverage and a soft, universal-fit nose bridge with cushion browguard for extra comfort and shock absorption.
1962.88 DKK - 2373.20 DKK
Specifications
Lens Type | DX |
---|---|
Certifications/Compliance | EN 166: EN 169 |
Product Type | Safety glasses |
For Use With (Application) | Construction |
Lens Material | Polycarbonate |
Description
- Safety spectacles with aspherical lens for 180° of Pure Vision and superior eye coverage
- DX coating provides enhanced anti-scratch and anti-fog protection
- Spectacles with ventilation channel across the lens to minimize misting
- Design provides excellent coverage and good field of vision
- Pliable, adjustable nose-pads and a 'soft-touch' brow guard
- Adjustable temple lengths and pantoscopic (angle) adjustments
Specifications
DX | |
Safety glasses | |
Polycarbonate |
EN 166: EN 169 | |
Construction |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
3M™ Maxim™ DX Safety Glasses