Learn More
Karl Hecht™ Mechanical Manual Counter
A four-figure counter with button for manual counting.
Brand: Karl Hecht™ 40344010
228.10 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Counter is held with the forefinger at the finger bow
- Counter is turned back to zero by using the lateral reset button
- Counts from 0 to 0000
- Casing made of chrome-plated steel
- Ergonomic design; lies comfortably in hand
Specifications
Manual Counter | |
0 to 9999 | |
Chrome-plated steel casing | |
Each |
Product Content Correction
Product Title
Karl Hecht™ Mechanical Manual Counter > Manual counter; 0 to 9999
Spot an opportunity for improvement?Share a Content Correction