Learn More
DWK Life Sciences Wheaton™ Media Lab Bottle, Non-Graduated, Amber
Click to view available options
Capacity (Metric):
125 mL
Description
- Manufactured from borosilicate glass which provides the best chemical resistance
- Amber borosilicate glass bottle provides protection from UV light
- Special pour lip facilitates pouring
- Packaged in partitioned carton to help prevent scratch marks on bottles
- Black phenolic screw cap can be purchased separately
Specifications
Specifications
Capacity (Metric) | 125 mL |
Color | Amber |
Material | Borosilicate Glass (Type I) |
Quantity | 48 EA |
Product Type | Media Bottle |
Closure Type | Not Included |