Learn More
OHAUS™ Medium Two Prong Sleeves
OHAUS™ Medium Two Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.
Brand: OHAUS™ 30400254
73.86 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Available in red vinyl and fiberglass
- Fiberglass sleeves are ideal for application above 100°C
Specifications
2 | |
Fiberglass | |
Gray |
Prong Sleeve | |
Medium |