Learn More
Mettler Toledo™ Membrane Kits
Brand: Mettler Toledo™ 51107692
4640.72 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Membrane Kit | |
Copper |
Promotions
-
Multiple Manufacturers
Focus On Clean Water
Shop now for up to 60% off Clean Water Research essentialsPromo Code: 24139Expires: 2025-08-21
New Offer Save Now
Product Content Correction
Product Title
Mettler Toledo™ Membrane Kits > Material: Copper
Spot an opportunity for improvement?Share a Content Correction