missing translation for 'onlineSavingsMsg'
Learn More
Menthofuran, 95%, Thermo Scientific Chemicals Product Code.: 11453407

Menthofuran, 95%, Thermo Scientific Chemicals

Product Code. 11453407
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11453407

Brand: Thermo Scientific Alfa Aesar A17192.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 494-90-6 | C10H14O | 150.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 494-90-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00041851
InChI Key YGWKXXYGDYYFJU-UHFFFAOYSA-N
Synonym menthofuran, menthofurane, 3,9-epoxy-p-mentha-3,8-diene, benzofuran, 4,5,6,7-tetrahydro-3,6-dimethyl, p-mentha-3,8-diene, 3,9-epoxy, 4,5,6,7-tetrahydro-3,6-dimethylbenzofuran, fema no. 3235, 4,5,6,7-tetrahydro-3,6-dimethylcoumarone, r-menthofuran, 3,6-dimethyl-4,5,6,7-tetrahydrobenzo 2,1-b furanShow MoreShow Less
PubChem CID 329983
ChEBI CHEBI:50542
IUPAC Name 3,6-dimethyl-4,5,6,7-tetrahydro-1-benzofuran
SMILES CC1CCC2=C(C1)OC=C2C

Specifications

Density 0.972
Boiling Point 86°C to 88°C (15 mmHg)
Flash Point 75°C (167°F)
Refractive Index 1.4845
Quantity 1 g
Formula Weight 150.22
Percent Purity 95%
Chemical Name or Material Menthofuran

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10244673 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberA17192.03,A17192.06
Lot Number10238989 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA17192.03,A17192.06
Lot Number10235780 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17192.03,A17192.06
Lot Number10107664 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17192.03,A17192.06
Lot NumberE2393B Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17192.03,A17192.06
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Menthofuran, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.