missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
Melting Point | 0°C |
Color | Colorless |
pH | <1 |
Physical Form | Liquid |
Quantity | 1000 mL |
Solubility Information | Soluble in Water |
Concentration or Composition (by Analyte or Components) | 0.05mol/L |
Odor | Odorless |
Safety and Handling


Product Identifier
- Mercury(II) nitrate 0.05 mol/l
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : mercury(II) nitrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mercury(II) Nitrate Solution. 0.05M, Chem Lab™ >
Spot an opportunity for improvement?Share a Content Correction