Learn More
Gonotec™ Mesasuring Beaker For Osmomat
Aids in the determination of total osmomality in aqueous solutions in very small sample volumes.
Brand: Gonotec™ 30.9.0010
This item is not returnable.
View return policy
Description
- Constantly controlled
- Expected reproducibility of the measuring results
- For use with the 3000/030/010 models
Specifications
1000/Pk. | |
Osmomat 010, 030, 3000 |
Product Content Correction
Product Title
Gonotec™ Mesasuring Beaker For Osmomat >
Spot an opportunity for improvement?Share a Content Correction