Learn More
MedChemExpress [Met5]-Enkephalin, amide TFA
MedChemExpress [Met5]-Enkephalin, amide TFA is an agonist for δ opioid receptors as well as putative ζ (zeta) opioid receptors.
Brand:  MedChemExpress HY-P1467A
Encompass_Preferred
Product Code. 16469548
  • 624.42 DKK / 10mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
98.35%
Neuroscience-Neuromodulation
Peptides
Room temperature
5-Methionine-enkephalin amide TFA
Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
686.7 g/mol
10 mg
H2O : 100 mg/mL (145.62 mM∣Need ultrasonic)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.