Learn More
S Murray™ Metal Luer-Lock Syringe
Use these syringes for a variety of research applications. S Murray™ Metal Luer-Lock Syringe are graduated and feature metal luer fittings.
Brand: S Murray™ G813/04
This item is not returnable.
View return policy
Description
Use these syringes for a variety of research applications. S Murray™ Metal Luer-Lock Syringe are graduated and feature metal luer fittings.
- Syringe with metal luer fitting
- Fully interchangeable
Specifications
CE | |
Metal Luer-lock Syringe | |
10 mL | |
Yes |
Glass | |
1/Ea. | |
Luer Lock |
Product Content Correction
Product Title
S Murray™ Metal Luer-Lock Syringe > Volume: 10mL
Spot an opportunity for improvement?Share a Content Correction