Learn More
Methoxychlor-d14 (bis(4-methoxyphenyl-d7)), Honeywell Fluka™ Product Code.: 15921131
Methoxychlor-d14 (bis(4-methoxyphenyl-d7)), Honeywell Fluka™
Product Code. 15921131 Shop All Honeywell Fluka Products
10 mg
Click to view available options
Quantity:
10 mg
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15921131
Brand: Honeywell Fluka R128610MG
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
CAS 1644449-82-0
Packaging Glass Bottle
Quantity 10 mg
For Use With (Application) Acaricides and Insecticides
Physical Form Powder
Grade Analytical Standard
Product Content Correction
Product Title
Methoxychlor-d14 (bis(4-methoxyphenyl-d7)), Honeywell Fluka™ >