missing translation for 'onlineSavingsMsg'
Learn More
Methyl 1-Boc-pyrrolidine-3-carboxylate, 97% Product Code.: 15466217

Methyl 1-Boc-pyrrolidine-3-carboxylate, 97%

Product Code. 15466217
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15466217

Brand: Thermo Scientific Alfa Aesar H33407.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122684-33-7 | C11H19NO4 | 229.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122684-33-7
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD04038683
InChI Key LIWFYAVKYUQMRE-UHFFFAOYSA-N
Synonym methyl 1-boc-3-pyrrolidinecarboxylate, 1-tert-butyl 3-methyl pyrrolidine-1,3-dicarboxylate, methyl 1-n-boc-dl-beta-prolinate, 1,3-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl 3-methyl ester, methyl 1-boc-3-pyrrolidine carboxylate, pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, methyl 1-boc-pyrrolidine-3-carboxylate, methyl 1-tert-butoxycarbonyl-3-pyrrolidinecarboxylate, s-1-boc-pyrrolidine-1,3-dicarboxylate, pubchem10748Show MoreShow Less
PubChem CID 4572775
IUPAC Name 1-O-tert-butyl 3-O-methyl pyrrolidine-1,3-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)C(=O)OC

Specifications

Boiling Point 105°C (0.2 mmHg)
Quantity 1 g
Formula Weight 229.28
Percent Purity 97%
Chemical Name or Material Methyl 1-Boc-pyrrolidine-3-carboxylate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10165321 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33407.06,H33407.03
Lot Number10224943 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33407.06,H33407.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 1-Boc-pyrrolidine-3-carboxylate, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.