Learn More
Methyl 2-formyl-4-methyl-4H-furo[3,2-b]pyrrole-5-carboxylate, 97%, Thermo Scientific™ Product Code.: 10653235
Methyl 2-formyl-4-methyl-4H-furo[3,2-b]pyrrole-5-carboxylate, 97%, Thermo Scientific™
Product Code. 10653235
Amber Glass Bottle; 1g
Click to view available options
Amber Glass Bottle; 1g
Product Code. 10653235
Brand: Thermo Scientific Maybridge MO07698DA
Specifications
CAS 164667-56-5
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H9NO4
MDL Number MFCD08060547
Quantity 1g
Synonym methyl 2-formyl-4-methyl-4h-furo 3,2-b pyrrole-5-carboxylate, methyl 2-formyl-4-methylfuro 3,2-b pyrrole-5-carboxylate, methyl 2-formyl-4-methylfurano 2,3-d pyrrole-5-carboxylate, 2-formyl-4-methyl-4h-furo 3,2-b pyrrole-5-carboxylic acid methyl ester
InChI Key VUQKFEPTRLQDRG-UHFFFAOYSA-N
SMILES CN1C(=CC2=C1C=C(O2)C=O)C(=O)OC
IUPAC Name methyl 2-formyl-4-methylfuro[3,2-b]pyrrole-5-carboxylate
Molecular Weight (g/mol) 207.185
PubChem CID 7537673
Formula Weight 207.18
Percent Purity 97%
Chemical Name or Material Methyl 2-formyl-4-methyl-4H-furo[3, 2-b]pyrrole-5-carboxylate
Show More Show Less
Product Content Correction
Product Title
Methyl 2-formyl-4-methyl-4H-furo[3,2-b]pyrrole-5-carboxylate, 97%, Thermo Scientific™ >